Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014911.1 | NP_001014911.1 | 2 - 3 | 342 | 113 | |
GenBank mRNA | BT021854.1 | - | 2 - 3 | 342 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |