Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BYSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014924.1 | NP_001014924.1 | 3 - 4 | 831 | 61 | |
GenBank mRNA | BC151370.1 | - | 3 - 4 | 884 | 61 | |
BT020890.1 | - | 3 - 4 | 831 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |