Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014939.1 | NP_001014939.1 | 3 - 4 | 396 | 141 | |
GenBank mRNA | BC102571.1 | - | 3 - 4 | 410 | 141 | |
BT021069.1 | - | 3 - 4 | 396 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |