Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014949.1 | NP_001014949.1 | 3 - 4 | 891 | 93 | |
GenBank mRNA | BC119880.1 | - | 3 - 4 | 946 | 93 | |
BT020824.1 | - | 3 - 4 | 891 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |