Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014950.1 | NP_001014950.1 | 2 - 3 | 432 | 99 | |
GenBank mRNA | BC102157.1 | - | 2 - 3 | 456 | 99 | |
BT021039.1 | - | 2 - 3 | 432 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |