Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014957.1 | NP_001014957.1 | 3 - 4 | 225 | 59 | |
GenBank mRNA | BC123510.1 | - | 2 - 3 | 190 | 59 | |
BT021513.1 | - | 3 - 4 | 225 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |