Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014967.1 | NP_001014967.1 | 2 - 3 | 224 | 87 | |
GenBank mRNA | BC140588.1 | - | 2 - 3 | 234 | 87 | |
BT021161.1 | - | 2 - 3 | 224 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |