Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015519.1 | NP_001015519.1 | 3 - 4 | 511 | 93 | |
GenBank mRNA | BC151482.1 | - | 3 - 4 | 502 | 93 | |
BT020860.1 | - | 3 - 4 | 511 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |