Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIBIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015541.1 | NP_001015541.1 | 1 | 3654 | 102 | |
GenBank mRNA | BT020949.1 | - | 1 | 3654 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |