Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015543.2 | NP_001015543.1 | 5 - 6 | 516 | 64 | |
GenBank mRNA | AY911336.1 | - | 5 - 6 | 517 | 64 | |
BC102865.1 | - | 5 - 6 | 520 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |