Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015585.1 | NP_001015585.1 | 7 - 8 | 718 | 61 | |
GenBank mRNA | BC111248.1 | - | 7 - 8 | 715 | 61 | |
BT021016.1 | - | 7 - 8 | 718 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |