Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015619.1 | NP_001015619.1 | 3 - 4 | 620 | 54 | |
GenBank mRNA | BC149811.1 | - | 3 - 4 | 615 | 54 | |
BT020661.1 | - | 3 - 4 | 517 | 54 | ||
BT020700.1 | - | 2 - 3 | 444 | 54 | ||
BT020802.1 | - | 2 - 3 | 411 | 54 | ||
BT020803.1 | - | 1 - 2 | 302 | 54 | ||
BT020853.1 | - | 3 - 4 | 620 | 54 | ||
BT020867.1 | - | 2 - 3 | 410 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |