Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MON1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015623.1 | NP_001015623.1 | 5 - 6 | 1661 | 63 | |
GenBank mRNA | BC118164.1 | - | 5 - 6 | 1718 | 63 | |
BT020883.1 | - | 5 - 6 | 1661 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |