Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDPGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015629.1 | NP_001015629.1 | 1 - 2 | 346 | 93 | |
GenBank mRNA | BT020839.1 | - | 1 - 2 | 346 | 93 | |
BT021160.1 | - | 1 - 2 | 344 | 93 | ||
BT021723.1 | - | 1 - 2 | 394 | 93 | ||
BT026292.1 | - | 1 - 2 | 310 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |