Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015650.1 | NP_001015650.1 | 8 - 9 | 1138 | 80 | |
GenBank mRNA | BC120337.1 | - | 8 - 9 | 1138 | 80 | |
BT020807.1 | - | 8 - 9 | 1138 | 80 | ||
BT026309.1 | - | 8 - 9 | 1127 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |