Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015658.1 | NP_001015658.1 | 1 - 2 | 168 | 80 | |
GenBank mRNA | BC109639.1 | - | 1 - 2 | 133 | 80 | |
BT020964.1 | - | 1 - 2 | 168 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |