Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015668.1 | NP_001015668.1 | 1 - 2 | 250 | 65 | |
GenBank mRNA | BC142022.1 | - | 1 - 2 | 330 | 65 | |
BT020842.1 | - | 1 - 2 | 250 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |