Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017951.1 | NP_001017951.1 | 1 - 2 | 507 | 65 | |
GenBank mRNA | BC114171.1 | - | 1 - 2 | 512 | 65 | |
BT021782.1 | - | 1 - 2 | 507 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |