Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017951.1 | NP_001017951.1 | 5 - 6 | 1191 | 62 | |
GenBank mRNA | BC114171.1 | - | 5 - 6 | 1196 | 62 | |
BT021782.1 | - | 5 - 6 | 1191 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |