Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017951.1 | NP_001017951.1 | 8 - 9 | 1429 | 69 | |
GenBank mRNA | BT021782.1 | - | 8 - 9 | 1429 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |