Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017952.1 | NP_001017952.1 | 3 - 4 | 388 | 98 | |
GenBank mRNA | BC110194.1 | - | 3 - 4 | 384 | 98 | |
BT020738.1 | - | 3 - 4 | 308 | 98 | ||
BT020771.1 | - | 3 - 4 | 396 | 98 | ||
BT020779.1 | - | 3 - 4 | 388 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |