Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024487.1 | NP_001019658.1 | 1 - 2 | 231 | 105 | |
GenBank mRNA | BC119955.1 | - | 1 - 2 | 183 | 105 | |
BT021763.1 | - | 1 - 2 | 231 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |