Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024516.1 | NP_001019687.1 | 3 - 4 | 690 | 123 | |
GenBank mRNA | BC120178.1 | - | 3 - 4 | 721 | 123 | |
BT021573.1 | - | 3 - 4 | 690 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |