Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024569.1 | NP_001019740.1 | 1 - 2 | 123 | 75 | |
GenBank mRNA | BC151526.1 | - | 1 - 2 | 109 | 75 | |
BT021517.1 | - | 1 - 2 | 123 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |