Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024569.1 | NP_001019740.1 | 6 - 7 | 801 | 77 | |
GenBank mRNA | BC151526.1 | - | 6 - 7 | 787 | 77 | |
BT021517.1 | - | 6 - 7 | 801 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |