Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024570.1 | NP_001019741.1 | 3 - 4 | 306 | 92 | |
GenBank mRNA | BC142215.1 | - | 3 - 4 | 336 | 92 | |
BT021849.1 | - | 3 - 4 | 306 | 92 | ||
BT030481.1 | - | 3 - 4 | 330 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |