Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025315.1 | NP_001020486.1 | 3 - 4 | 333 | 102 | |
GenBank mRNA | AY911354.1 | - | 3 - 4 | 333 | 102 | |
BC102560.1 | - | 1 | 334 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |