Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025315.1 | NP_001020486.1 | 4 - 5 | 443 | 121 | |
GenBank mRNA | AY911354.1 | - | 4 - 5 | 443 | 121 | |
BC102560.1 | - | 1 | 444 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |