Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025329.2 | NP_001020500.1 | 4 - 5 | 418 | 120 | |
GenBank mRNA | AY911362.1 | - | 4 - 5 | 419 | 120 | |
BC109650.1 | - | 4 - 5 | 477 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |