Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025339.2 | NP_001020510.1 | 5 - 6 | 454 | 87 | |
GenBank mRNA | AY911383.1 | - | 5 - 6 | 455 | 87 | |
BC103133.1 | - | 6 - 7 | 456 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |