Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025569.1 | NP_001020740.1 | 1 - 2 | 471 | 70 | |
GenBank mRNA | AB605262.1 | - | 1 - 2 | 400 | 70 | |
BC102129.1 | - | 1 - 2 | 455 | 70 | ||
BT021550.1 | - | 1 - 2 | 474 | 70 | ||
CR850362.2 | - | 1 - 2 | 471 | 70 | ||
KX806593.1 | - | 1 - 2 | 350 | 70 | ||
KX806594.1 | - | 1 - 2 | 350 | 70 | ||
KX806595.1 | - | 1 - 2 | 350 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |