Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033120.1 | NP_001028292.1 | 3 - 4 | 354 | 99 | |
GenBank mRNA | AB205159.1 | - | 3 - 4 | 354 | 99 | |
BC149398.1 | - | 3 - 4 | 557 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |