Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034209.1 | NP_001029381.1 | 1 - 2 | 793 | 61 | |
GenBank mRNA | BT020794.1 | - | 1 - 2 | 793 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |