Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034231.2 | NP_001029403.1 | 2 - 3 | 553 | 66 | |
GenBank mRNA | BC102921.1 | - | 2 - 3 | 555 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |