Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034264.1 | NP_001029436.1 | 1 - 2 | 284 | 151 | |
GenBank mRNA | BC103308.1 | - | 1 - 2 | 284 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |