Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034331.1 | NP_001029503.1 | 3 - 4 | 660 | 69 | |
GenBank mRNA | BC102237.1 | - | 3 - 4 | 660 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |