Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034364.2 | NP_001029536.1 | 3 - 4 | 420 | 54 | |
GenBank mRNA | BC102511.1 | - | 3 - 4 | 421 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |