Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034383.1 | NP_001029555.1 | 1 - 2 | 49 | 85 | |
GenBank mRNA | BC103058.1 | - | 1 - 2 | 49 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Inventoried Probe spans exons |