Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034507.2 | NP_001029679.1 | 1 - 2 | 70 | 104 | |
GenBank mRNA | BC102506.1 | - | 1 - 2 | 71 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |