Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034534.1 | NP_001029706.1 | 1 - 2 | 287 | 88 | |
GenBank mRNA | BC103222.1 | - | 1 - 2 | 287 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |