Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034537.1 | NP_001029709.1 | 2 - 3 | 357 | 81 | |
GenBank mRNA | BC119916.1 | - | 2 - 3 | 341 | 81 | |
BT020805.1 | - | 2 - 3 | 357 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |