Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034678.1 | NP_001029850.1 | 3 - 4 | 513 | 67 | |
GenBank mRNA | BC103309.1 | - | 3 - 4 | 513 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |