Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCHF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034715.2 | NP_001029887.1 | 5 - 6 | 1044 | 148 | |
GenBank mRNA | BC102966.1 | - | 5 - 6 | 1046 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |