Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034811.1 | NP_001029983.1 | 1 - 2 | 239 | 66 | |
GenBank mRNA | BC103476.1 | - | 1 - 2 | 54 | 66 | |
BT021724.1 | - | 1 - 2 | 239 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |