Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMUR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035048.1 | NP_001030220.1 | 1 - 2 | 827 | 79 | |
GenBank mRNA | BT021833.1 | - | 1 - 2 | 827 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Bos taurus Probe spans exons |