Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035075.1 | NP_001030247.1 | 1 - 2 | 102 | 80 | |
GenBank mRNA | BT021626.1 | - | 1 - 2 | 102 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |