Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035082.1 | NP_001030254.1 | 8 - 9 | 1019 | 115 | |
GenBank mRNA | BT021529.1 | - | 8 - 9 | 1019 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |