Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK2AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035320.1 | NP_001030397.1 | 3 - 4 | 368 | 74 | |
GenBank mRNA | BT021910.1 | - | 3 - 4 | 368 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |