Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035330.2 | NP_001030407.1 | 2 - 3 | 263 | 124 | |
GenBank mRNA | BC102517.1 | - | 2 - 3 | 266 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |