Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035450.1 | NP_001030527.1 | 4 - 5 | 383 | 152 | |
GenBank mRNA | BC119872.1 | - | 4 - 5 | 418 | 152 | |
BT020837.1 | - | 4 - 5 | 383 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |